Anti-OR51A4 Rabbit Polyclonal Antibody
ANTIA99527-100
New Product
- Antibody type:Primary
- Antigen name:O51A4_HUMAN
- Antigen symbol:OR51A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8NGJ6
- Antigen synonyms:Olfactory receptor 51A4|Olfactory receptor, family 51, subfamily A, member 4|OR51A4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR51A4 (amino acids 200-249)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to OR51A4 for WB, IF and ELISA with samples derived from human.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: OR51A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human