Anti-alpha 1d Adrenergic Receptor Rabbit Polyclonal Antibody
Catalog # ANTIA99322-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ADA1D_HUMAN
- Antigen symbol:alpha 1d Adrenergic Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P25100
- Antigen synonyms:Adra1d|Adrenergic receptor alpha 1d|Adrenergic receptor delta1|Adrenergic alpha1A receptor|Adra1a|Adra1|ADRA1R|Adrd1|Adrenergic alpha1D receptor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ADRA1D (amino acids 451-500)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ADRA1D for IHC, IF and ELISA with samples derived from human.
- Validated applications: IHC, IF, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: alpha 1d Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human