Order Entry
Export
ContactUsLinkComponent
 
Anti-C9ORF46 Rabbit Polyclonal Antibody
Catalog # ANTIA99273-100
Supplier:  Antibodies.com
New Product
undefined
Anti-C9ORF46 Rabbit Polyclonal Antibody
Catalog # ANTIA99273-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AD025
  • Antigen symbol:
    C9ORF46
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9HBL7
  • Antigen synonyms:
    Hypothetical protein LOC55848|FLJ14688|Uncharacterized hematopoietic stem/progenitor cells protein MDS030|Chromosome 9 open reading frame 46|FLJ39176|transmembrane protein C9orf46|C9ORF46|MDS030
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    17 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from the Internal region of human PLGRKT (amino acids 1-50)
  • Purification:
    The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to PLGRKT for WB and ELISA with samples derived from human and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:2000, ELISA: 1:10000

Type: Primary
Antigen: C9ORF46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat