Anti-Cytokeratin 8+18 Rabbit Polyclonal Antibody
Catalog # ANTIA99240-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CARD2
- Antigen symbol:Cytokeratin 8+18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05787
- Antigen synonyms:CYK8|CK-8|CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|CK 8|Cell proliferation inducing gene 46 protein|CK8
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human K8 around the non-acetylation site of Lys483 (amino acids 434-483)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to K8 (Ab-483) for WB and ELISA with samples derived from human.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Cytokeratin 8+18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human