Anti-HOXD10 Rabbit Polyclonal Antibody
Catalog # ANTIA98803-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AI385591
- Antigen symbol:HOXD10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P28358
- Antigen synonyms:Homeo box 4D|Homeobox protein Hox-D10|Hox 4.5|Homeobox D10|Homeo box D10|Homeobox protein Hox-4D|Homeobox protein Hox-4E|Hox 4.4|AI874987
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human HOXD10 (amino acids 291-340)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to HOXD10 for WB and ELISA with samples derived from human and mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: HOXD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse