Anti-ZFP106 Rabbit Polyclonal Antibody
ANTIA98732-100
New Product
- Antibody type:Primary
- Antigen name:DKFZp451A239
- Antigen symbol:ZFP106
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9H2Y7
- Antigen synonyms:Zinc finger protein 106 homolog|SH3 domain binding protein 3|FLJ45841|Zfp 106|Zinc finger protein 106 homolog (mouse)|FLJ34610|ZFP106|SH3BP3|Zinc finger protein 474
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:208 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ZFP106 (amino acids 1833-1882)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to ZFP106 for WB and ELISA with samples derived from human and mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ZFP106
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse