Anti-TRP2/DCT Rabbit Polyclonal Antibody
Catalog # ANTIA98674-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:L dopachrome tautomerase
- Antigen symbol:TRP2/DCT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P40126
- Antigen synonyms:EC 5.3.3.12|TRP 2|Dopachrome delta isomerase|DT|L-dopachrome tautomerase|L dopachrome Delta isomerase|Dopachrome tautomerase dopachrome delta isomerase, tyrosine related protein 2|DCT|L-dopachrome Delta-isomerase
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human DCT (amino acids 51-100)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to DCT for WB, IF and ELISA with samples derived from human and mouse.
- Validated applications: WB, IF, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: TRP2/DCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse