Anti-Nesprin3 Rabbit Polyclonal Antibody
Catalog # ANTIA98361-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:C14orf49
- Antigen symbol:Nesprin3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q6ZMZ3
- Antigen synonyms:Nesprin-3|NET53|Nesp3|MGC75267|Nuclear Envelope Spectrin Repeat Protein 3|Nesprin 3|Chromosome 14 open reading frame 49|FLJ25605|Nesprin3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:112 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human C14orf49 (amino acids 169-218)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to C14orf49 for WB and ELISA with samples derived from human and mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: Nesprin3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse