Anti-Bcl2L2 Rabbit Polyclonal Antibody
Catalog # ANTIA98270-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Apoptosis regulator BCL W
- Antigen symbol:Bcl2L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q92843
- Antigen synonyms:Bcl2 L2|BCL2 like 2 protein|BCL 2 Like 2|BCL2 like 2|Bcl-2-like protein 2|Bcl 2L2|Bcl 2 like 2 protein|Apoptosis regulator Bcl-W|B2CL2_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BCLW (amino acids 105-154)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to BCLW for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Bcl2L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat