Anti-BIN1 Rabbit Polyclonal Antibody
Catalog # ANTIA98267-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AMPH 2
- Antigen symbol:BIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O00499
- Antigen synonyms:BIN1|amphiphysin-like|Amphiphysin-like protein|Amphiphysin II|Amphiphysin like protein|AMPH2|BIN1_HUMAN|Amphiphysin 2|AMPHL
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:64 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the C-terminal region of human BIN1 (amino acids 521-570)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to BIN1 for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: BIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat