Anti-CCR4 Rabbit Polyclonal Antibody
Catalog # ANTIA98255-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:C C chemokine receptor type 4
- Antigen symbol:CCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P51679
- Antigen synonyms:CCR4_HUMAN|CC CKR 4|CCR-4|CC-CKR-4|C-C chemokine receptor type 4|CCR4|C-C CKR-4|C C CKR 4|CCR 4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the N-terminal region of human CCR4 (amino acids 1-50)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to CCR4 for WB and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: CCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat