Order Entry
Export
ContactUsLinkComponent
Anti-TCP1 eta Rabbit Polyclonal Antibody
Anti-TCP1 eta Rabbit Polyclonal Antibody
Catalog # ANTIA98251-100
Supplier:  Antibodies.com
New Product
undefined
Anti-TCP1 eta Rabbit Polyclonal Antibody
Catalog # ANTIA98251-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CCT ETA
  • Antigen symbol:
    TCP1 eta
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q99832
  • Antigen synonyms:
    HIV 1 Nef interacting protein|Chaperonin containing t complex polypeptide 1 eta subunit|CCT7|Ccth|Chaperonin containing TCP1 subunit 7 (eta)|CCTETA|CCT-eta|HIV-1 Nef-interacting protein|Chaperonin containing TCP1 subunit 7
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    59 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human CCT7 (amino acids 209-258)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to CCT7 for WB and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: TCP1 eta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat