Order Entry
Export
ContactUsLinkComponent
Anti-MUC16 Mouse Monoclonal Antibody [clone: ABT-MUC16]
Anti-MUC16 Mouse Monoclonal Antibody [clone: ABT-MUC16]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-MUC16 Mouse Monoclonal Antibody [clone: ABT-MUC16]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA99327-50
  • ANTIA99327-100
  • Antigen symbol:
  • MUC16
  • MUC16
  • Antigen name:
  • CA 125
  • CA 125
  • Antigen synonyms:
  • MUC 16|MUC-16|CA-125|CA125 ovarian cancer antigen|MUC16_HUMAN|Cancer antigen 125|MUC16|FLJ14303|CA125
  • MUC 16|MUC-16|CA-125|CA125 ovarian cancer antigen|MUC16_HUMAN|Cancer antigen 125|MUC16|FLJ14303|CA125
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-MUC16
  • ABT-MUC16
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# Q8WXI7
  • UniprotID# Q8WXI7
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide derived from human CA-125
  • Synthetic peptide derived from human CA-125
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC
  • IHC

Specifications