Order Entry
Export
ContactUsLinkComponent
 
Anti-WIPI1 Rabbit Monoclonal Antibody [clone: ARC1652]
  ANTIA307185-100
New Product
undefined
Anti-WIPI1 Rabbit Monoclonal Antibody [clone: ARC1652]
  ANTIA307185-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ATG 18
  • Antigen symbol:
    WIPI1
  • Clonality:
    Monoclonal
  • Clone:
    ARC1652
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q5MNZ9
  • Antigen synonyms:
    ATG18A|WD repeat domain phosphoinositide interacting protein 1|FLJ10055|WD40 repeat protein interacting with phosphoinositides of 49 kDa|Atg18 protein homolog|WD repeat domain phosphoinositide interacting 1|ATG18|WD repeat domain phosphoinositide-interacting protein 1|WD40 repeat protein interacting with phosphoInositides of 49kDa
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    49 kDa
  • Sequence:
    MEAEAADAPPGGVESALSCFSFNQDCTSLATGTKAGYKLFSLSSVEQLDQVHGSNEIPDVYIVERLFSSSLVVVVSHTKPRQMNVYHFKKGTEICNYSYS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1-100 of human WIPI1 (Q5MNZ9).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC1652] antibody to WIPI1 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: WIPI1
Clonality: Monoclonal
Clone: ARC1652
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat