Anti-GilZ/TilZ Rabbit Polyclonal Antibody
ANTIA307146-100
New Product
- Antibody type:Primary
- Antigen name:Delta sleep-inducing peptide immunoreactor
- Antigen symbol:GilZ / TilZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q99576
- Antigen synonyms:Dsipi|GilZ / TilZ|GILZ|DSIP-immunoreactive peptide|Glucocorticoid-induced leucine zipper protein|hDIP|DIP|Dsip1|DSIP immunoreactive leucine zipper protein
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
- Molecular weight:16 kDa
- Sequence:MDLVKNHLMYAVREEVEILKEQIRELVEKNSQLERENTLLKTLASPEQLEKFQSCLSPEEPAPESPQVPEAPGGSAV
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-79 of human TSC22D3 (NP_001015881.1).
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to GilZ/TilZ for WB, IHC, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF, IP
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200, IP: 1:50 to 1:100
Type: Primary
Antigen: GilZ / TilZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat