Anti-BBS7 Rabbit Polyclonal Antibody
Catalog # ANTIA307144-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bardet Biedl syndrome 7 protein
- Antigen symbol:BBS7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8IWZ6
- Antigen synonyms:BBS2L1|Bardet-Biedl syndrome 7|BBS2-like protein 1|BBS2 like protein 1|FLJ10715|Bardet-Biedl syndrome 7 protein|BBS7|BBS7_HUMAN|BBS2 like 1
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
- Molecular weight:80 kDa
- Sequence:MDLILNRMDYLQVGVTSQKTMKLIPASRHRATQKVVIGDHDGVVMCFGMKKGEAAAVFKTLPGPKIARLELGGVINTPQEKIFIAAASEIRGFTKRGKQFLSFETNLTESIKAMHISGSDLFLSASYIYNHYCDCKDQHYYLSGDKINDVICLPVERLSRITPVLACQDRVLRVLQGSDVMYAVEVPGPPTVLALHNGNGGDSGEDLLFGTSDGKLALIQITTSKPVRKWEIQNEKKRGGILCIDSFDIVGDGVKDLLVGRDDGMVEVYS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human BBS7 (NP_060660.2).
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to BBS7 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:100, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: BBS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat