Order Entry
Export
ContactUsLinkComponent
Anti-p53 (acetyl Lys370) Rabbit Monoclonal Antibody [clone: ARC2368]
Anti-p53 (acetyl Lys370) Rabbit Monoclonal Antibody [clone: ARC2368]
Catalog # ANTIA306200-100
Supplier:  Antibodies.com
New Product
undefined
Anti-p53 (acetyl Lys370) Rabbit Monoclonal Antibody [clone: ARC2368]
Catalog # ANTIA306200-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen NY-CO-13
  • Antigen symbol:
    p53
  • Clonality:
    Monoclonal
  • Clone:
    ARC2368
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04637
  • Antigen synonyms:
    P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
  • Modification:
    acetyl Lys370
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    53 kDa
  • Sequence:
    HLKSK
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic acetylated peptide around K370 of human p53 (P04637).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal antibody to p53 (acetyl Lys370) for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: ARC2368
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat