Order Entry
Export
ContactUsLinkComponent
Anti-Histone H4 Rabbit Polyclonal Antibody
Anti-Histone H4 Rabbit Polyclonal Antibody
Catalog # ANTIA306062-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Histone H4 Rabbit Polyclonal Antibody
Catalog # ANTIA306062-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dJ160A22.1
  • Antigen symbol:
    Histone H4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P62805
  • Antigen synonyms:
    FO108|H4|dJ160A22.2|dJ221C16.9|H4 histone family, member D|dJ221C16.1|H4 histone family, member B|H4 histone family, member C|H4 histone family, member A
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    11 kDa
  • Sequence:
    GLGKG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic acetylated peptide around K12 of human Histone H4 (NP_001029249.1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Histone H4 for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: Histone H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat