Order Entry
Export
ContactUsLinkComponent
Anti-IL-18 Rabbit Polyclonal Antibody
Anti-IL-18 Rabbit Polyclonal Antibody
Catalog # ANTIA308037-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-IL-18 Rabbit Polyclonal Antibody
Catalog # ANTIA308037-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Iboctadekin
  • Antigen symbol:
    IL-18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P70380
  • Antigen synonyms:
    IFN gamma inducing factor|IL-1 gamma|IL 1g|IL 18|IFN-gamma-inducing factor|IGIF|IL1 gamma|IL-18|IL 1 gamma
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
  • Molecular weight:
    22 kDa/18 kDa
  • Sequence:
    NFGRLHCTTAVIRNINDQVLFVDKRQPVFEDMTDIDQSASEPQTRLIIYMYKDSEVRGLAVTLSVKDSKMSTLSCKNKIISFEEMDPPENIDDIQSDLIFFQKRVPGHNKMEFESSLYEGHFLACQKEDDAFKLILKKKDENGDKSVMFTLTNLHQS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 36-192 of mouse IL18 (NP_032386.1)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to IL-18 for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200

Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat