Anti-EGFR Rabbit Recombinant Antibody [clone: DM128]
New Product
Recombinant rabbit monoclonal [DM128] antibody to EGFR for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: EGFR
Clonality: Recombinant
Clone: DM128
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ANTIA318579-500
- ANTIA318579-100
- Antigen symbol:
- EGFR
- EGFR
- Antigen name:
- 9030024J15Rik
- 9030024J15Rik
- Antigen synonyms:
- EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
- EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM128
- DM128
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P00533
- UniprotID# P00533
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C