Anti-SLC31A1 / CTR1 Rabbit Monoclonal Antibody [clone: ARC2537]
ANTIA307135-100
New Product
- Antibody type:Primary
- Antigen name:Copper transport 1 homolog
- Antigen symbol:SLC31A1 / CTR1
- Clonality:Monoclonal
- Clone:ARC2537
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O15431
- Antigen synonyms:hCTR1|COPT1_HUMAN|High affinity copper uptake protein 1|SLC31A1 / CTR1|Copper transporter 1|COPT1|SLC31A1|CTR1|solute carrier family 31 (copper tansporters) member 1
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molecular weight:32 kDa
- Sequence:MDHSHHMGMSYMDSNSTMQPSHHHPTTSASHSHGGGDSSMMMMPMTFYFGFKNVELLFSGLVINTAGEMAGAFVAVFLLAMFYEGLKIARESLLRKSQVS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CTR1/SLC31A1 (O15431).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit monoclonal [ARC2537] antibody to SLC31A1 / CTR1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1000
Type: Primary
Antigen: SLC31A1 / CTR1
Clonality: Monoclonal
Clone: ARC2537
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat