Order Entry
Export
ContactUsLinkComponent
Anti-RBBP4 Rabbit Monoclonal Antibody [clone: ARC0813]
Anti-RBBP4 Rabbit Monoclonal Antibody [clone: ARC0813]
  ANTIA306852-100
New Product
undefined
Anti-RBBP4 Rabbit Monoclonal Antibody [clone: ARC0813]
  ANTIA306852-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CAF I p48
  • Antigen symbol:
    RBBP4
  • Clonality:
    Monoclonal
  • Clone:
    ARC0813
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q09028
  • Antigen synonyms:
    Chromatin assembly factor I p48 subunit|Nucleosome-remodeling factor subunit RBAP48|MSI1 protein homolog|CAF-I p48|CAF-I 48 kDa subunit|Chromatin assembly factor 1 subunit C|Histone-binding protein RBBP4|Chromatin assembly factor/CAF 1 p48 subunit|CAF-1 subunit C
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    52 kDa
  • Sequence:
    MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RBBP4 (Q09028).
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC0813] antibody to RBBP4 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: RBBP4
Clonality: Monoclonal
Clone: ARC0813
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat