Order Entry
Export
ContactUsLinkComponent
Anti-V5 Tag Rabbit Polyclonal Antibody
Anti-V5 Tag Rabbit Polyclonal Antibody
Catalog # ANTIA306052-100
Supplier:  Antibodies.com
New Product
undefined
Anti-V5 Tag Rabbit Polyclonal Antibody
Catalog # ANTIA306052-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GKPIPNPLLGLDST epitope tag
  • Antigen symbol:
    V5 Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Species independent
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    V5 epitope tag|V5 tag|Regulator of expression of viral proteins|Protein Rev|GKPIPNPLLGLDST tag
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Sequence:
    GKPIPNPLLGLDST
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to V5 tag.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to V5 Tag for WB with samples derived from Species Independent.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:1000 to 1:2000

Type: Primary
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Species Independent