Anti-ARPC1B Goat Polyclonal Antibody
Catalog # ANTIA83839-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Actin related protein 2/3 complex subunit 1B
- Antigen symbol:ARPC1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ARC1B_HUMAN|Arpc1b|Actin-related protein 2/3 complex subunit 1B|Arp2/3 complex 41 kDa subunit|ARC41|ARP2/3 subunit 1B|ARP2/3 protein complex subunit p41|Actin related protein 2/3 complex subunit 1B 41kDa|p40 ARC
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:AYHSFLVEPISCH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ARPC1B (N terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to ARPC1B for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: ARPC1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human