Anti-SH3GLB1 Goat Polyclonal Antibody
Catalog # ANTIA83782-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bax interacting factor 1
- Antigen symbol:SH3GLB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Bif 1|Bif-1|CGI 61|Endophilin B1|PPP1R70|Endophilin-B1|Bax-interacting factor 1|dJ612B15.2|Bif
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-QKGKVPITYLELLN
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human SH3GLB1 (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to SH3GLB1 for ELISA, WB, IHC and flow cytometry with samples derived from human, mouse and rat.
- Validated applications: ELISA, WB, IHC, flow cytometry
Type: Primary
Antigen: SH3GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat