Anti-Vinculin Rabbit Recombinant Antibody [clone: VCL/8533R]
Catalog # ANTIA317168-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CMD1W
- Antigen symbol:Vinculin
- Clonality:Recombinant
- Clone:VCL/8533R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P18206
- Antigen synonyms:MVCL|CMH15|OTTHUMP00000019861|VINC|HEL114|MV|Epididymis luminal protein 114|Metavinculin|OTTHUMP00000019862
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
- Molecular weight:117 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 174-322) of human Vinculin (VCL) protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [VCL/8533R] antibody to Vinculin for WB and IHC-P with samples derived from human and mouse.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Vinculin
Clonality: Recombinant
Clone: VCL/8533R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human;Mouse