Anti-TrkB Mouse Monoclonal Antibody [clone: NTRK2/7926]
ANTIA317134-100
New Product
- Antibody type:Primary
- Antigen name:AI848316
- Antigen symbol:TrkB
- Clonality:Monoclonal
- Clone:NTRK2/7926
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2c lambda
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q16620
- Antigen synonyms:GP145-TrkB/GP95-TrkB|BDNF/NT-3 growth factors receptor|EC 2.7.10.1|BDNF/NT 3 growth factors receptor|GP145-TrkB|GP145 TrkB|C030027L06Rik|BDNF tropomyosine receptor kinase B|Brain derived neurotrophic factor receptor
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without Sodium azide and carrier free
- Molecular weight:95 - 145 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 250-450) of human NTRK2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [NTRK2/7926] antibody to TrkB for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: TrkB
Clonality: Monoclonal
Clone: NTRK2/7926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c lambda
Reactivity: Human