Anti-HSPA8 Goat Polyclonal Antibody
Catalog # ANTIA83237-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:2410008N15Rik
- Antigen symbol:HSPA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Epididymis luminal protein 33|Heat shock 70kDa protein 8|Heat shock cognate protein 54|Epididymis secretory sperm binding protein Li 72p|Heat shock 70kD protein 8|Heat shock 70 kDa protein 8|Heat shock 70kD protein 10|Heat shock cognate 71 kDa protein|Constitutive heat shock protein 70
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-EKLQGKINDEDKQK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human HSPA8 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to HSPA8 for ELISA, WB, IF and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IF, IHC
Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse