Anti-IDS Goat Polyclonal Antibody
Catalog # ANTIA83156-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Alpha L iduronate sulfate sulfatase
- Antigen symbol:IDS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Alpha-L-iduronate sulfate sulfatase|Iduronate 2 sulfatase 42 kDa chain|AW214631|Iduronate sulfatase|IDS_HUMAN|Iduronate 2 sulfatase 14 kDa chain|Iduronate 2-sulfatase 14 kDa chain|Iduronate 2 sulfatase|Ids
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-KHFRFRDLEEDP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human IDS (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to IDS for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: IDS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human