Anti-NQO1 Goat Polyclonal Antibody
Catalog # ANTIA82564-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Azoreductase
- Antigen symbol:NQO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Diaphorase 4|Diaphorase (NADH/NADPH) (cytochrome b 5 reductase)|Dioxin inducible 1|Diaphorase (NADH/NADPH)|DIA4|DT diaphorase|DHQU|DIA 4|Cytochrome b 5 reductase
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-DKGPFRSKKAVLS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human NQO1 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to NQO1 for ELISA, WB, IF and IHC with samples derived from human.
- Validated applications: ELISA, WB, IF, IHC
Type: Primary
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human