Anti-Amyloid Precursor Protein Rabbit Polyclonal Antibody
Catalog # ANTIA451-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:A4 amyloid protein
- Antigen symbol:Amyloid Precursor Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05067
- Antigen synonyms:AICD-59|ABETA|AD1|A4_HUMAN|ABPP|AID(50)|AICD-50|AICD-57|AAA
- Storage buffer:Supplied as antiserum with 0,05% Sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C-terminus of the caspase 3-cleaved human APP protein
- Tested applications:ICC
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to APP delta C31 for WB, ICC and ELISA with samples derived from human, mouse and rat.
- Validated applications: WB, ICC, ELISA
- Recommended dilutions: WB: 1:1000 to 1:3000, ICC: 1:500 to 1:1000
Type: Primary
Antigen: Amyloid Precursor Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat