Order Entry
Export
ContactUsLinkComponent
Anti-p53 Mouse Monoclonal Antibody [clone: 2B7E4]
Anti-p53 Mouse Monoclonal Antibody [clone: 2B7E4]
Catalog # ANTIA404-100
Supplier:  Antibodies.com
New Product
undefined
Anti-p53 Mouse Monoclonal Antibody [clone: 2B7E4]
Catalog # ANTIA404-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen NY-CO-13
  • Antigen symbol:
    p53
  • Clonality:
    Monoclonal
  • Clone:
    2B7E4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04637
  • Antigen synonyms:
    P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% Glycerol
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide containing acetyl Lys382 of human p53 protein
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal (2B7E4) antibody to p53 (acetyl K382) for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1 µg/ml

Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: 2B7E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human