Anti-p53 Mouse Monoclonal Antibody [clone: #18]
ANTIA402-100
New Product
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Monoclonal
- Clone:#18
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% Glycerol (filter-sterilised, without Sodium azide and carrier free)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide containing phospho Ser315 of human p53 protein
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal (#18) antibody to p53 (phospho Ser315) for WB, IHC, IF and ELISA with samples derived from human.
- Validated applications: WB, IHC, IF, ELISA
- Recommended dilutions: WB: 1:1000 to 1:2000
Type: Primary
Antigen: p53
Clonality: Monoclonal
Clone: #18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human