Order Entry
Export
ContactUsLinkComponent
Anti-EGFR Recombinant Recombinant Antibody
Anti-EGFR Recombinant Recombinant Antibody
  ANTIA318922-100
New Product
undefined
Anti-EGFR Recombinant Recombinant Antibody
  ANTIA318922-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    9030024J15Rik
  • Antigen symbol:
    EGFR
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Recombinant
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Lyophilized
  • Gene ID:
    UniprotID# P00533
  • Antigen synonyms:
    EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
  • Storage buffer:
    Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
    Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Concentration:
    Reconstitution dependent
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant chimeric monoclonal [Cetuximab Biosimilar] antibody to EGFR for ELISA with samples derived from human.

  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1:5000 to 10000

Type: Primary
Antigen: EGFR
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human