Order Entry
Export
ContactUsLinkComponent
 
Anti-IL-20 Human Recombinant Antibody
  ANTIA318838-100
New Product
undefined
Anti-IL-20 Human Recombinant Antibody
  ANTIA318838-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    IL-20
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Human
  • Isotype:
    IgG4
  • Reactivity:
    Human
  • Form:
    Lyophilized
  • Gene ID:
    UniprotID# Q9NYY1
  • Storage buffer:
    Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
    Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Concentration:
    Reconstitution dependent
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant human monoclonal [Fletikumab Biosimilar] antibody to IL-20 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100

Type: Primary
Antigen: IL-20
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG4
Reactivity: Human