Anti-CD44 Recombinant Recombinant Antibody [clone: DMC269]
New Product
Recombinant chimeric monoclonal [DMC269] antibody to CD44 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CD44
Clonality: Recombinant
Clone: DMC269
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
- Catalog No:
- ANTIA318789-500
- ANTIA318789-100
- Antigen symbol:
- CD44
- CD44
- Antigen name:
- BA-1
- BA-1
- Antigen synonyms:
- CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
- CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC269
- DMC269
- Reactivity:
- Human
- Human
- Host:
- Recombinant
- Recombinant
- Gene ID:
- UniprotID# P16070
- UniprotID# P16070
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C