Anti-Albumin Recombinant Recombinant Antibody [clone: DMC275]
Supplier: Antibodies.com
New Product
Certificates
About this item
Recombinant chimeric monoclonal [DMC275] antibody to Albumin for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: Albumin
Clonality: Recombinant
Clone: DMC275
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ANTIA318783-500
- ANTIA318783-100
- Antigen symbol:
- Albumin
- Albumin
- Antigen name:
- Albumin
- Albumin
- Antigen synonyms:
- GIG20|DKFZp779N1935|ALBU_HUMAN|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Growth inhibiting protein 20|Albumin
- GIG20|DKFZp779N1935|ALBU_HUMAN|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Growth inhibiting protein 20|Albumin
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC275
- DMC275
- Reactivity:
- Human
- Human
- Host:
- Recombinant
- Recombinant
- Gene ID:
- UniprotID# P02768
- UniprotID# P02768
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C