Anti-S100A1 Mouse Monoclonal Antibody [clone: S1/61]
ANTIA253097-100
New Product
- Antibody type:Primary
- Antigen symbol:S100A1
- Clonality:Monoclonal
- Clone:S1/61
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P23297
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:10 - 12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified human S-100 protein conjugated to methylated BSA
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S1/61] antibody to S100A1 for IF and IHC-P with samples derived from Human.
- Validated applications: IF, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: S100A1
Clonality: Monoclonal
Clone: S1/61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human