Anti-ACTH Mouse Monoclonal Antibody [clone: AH26 + 57]
Catalog # ANTIA252841-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ACTH
- Antigen symbol:ACTH
- Clonality:Monoclonal
- Clone:AH26 + 57
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P01189
- Antigen synonyms:Beta melanocyte stimulating hormone|Beta LPH|alphaMSH|Beta-endorphin|Adrenocorticotropic hormone|alpha-MSH|Alpha melanocyte stimulating hormone|Adrenocorticotropin|beta-MSH
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:ACTH is ~5 kDa/the POMC precursor is ~30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone AH26: Synthetic peptide corresponding to amino acids 1 - 24 of human ACTH protein. Clone 57: N terminal fragment of human ACTH protein, conjugated to KLH
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [AH26 + 57] antibody to ACTH for IHC-P with samples derived from Human, Mouse and Rat.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: ACTH
Clonality: Monoclonal
Clone: AH26 + 57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human;Mouse;Rat