Order Entry
Export
ContactUsLinkComponent
Anti-c-Myc Mouse Monoclonal Antibody [clone: CT14.G4]
Anti-c-Myc Mouse Monoclonal Antibody [clone: CT14.G4]
Catalog # ANTIA252629-100
Supplier:  Antibodies.com
New Product
Anti-c-Myc Mouse Monoclonal Antibody [clone: CT14.G4]
Catalog # ANTIA252629-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AU016757
  • Antigen symbol:
    c-Myc
  • Clonality:
    Monoclonal
  • Clone:
    CT14.G4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Gorilla,
    Chimpanzee,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01106
  • Antigen synonyms:
    MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    62 - 64 kDa
  • Sequence:
    AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to amino acids 408 - 439 of human c - Myc, coupled to KLH
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CT14.G4] antibody to c-Myc for Flow Cytometry and IF with samples derived from Human, Chimpanzee and Gorilla.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: CT14.G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Chimpanzee;Gorilla