Order Entry
Export
ContactUsLinkComponent
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/2597]
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/2597]
Catalog # ANTIA252263-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: ITGB3/2597]
Catalog # ANTIA252263-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BDPLT16
  • Antigen symbol:
    Integrin beta 3
  • Clonality:
    Monoclonal
  • Clone:
    ITGB3/2597
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05106
  • Antigen synonyms:
    HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    105/90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 385 - 490, of human ITGB3 protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [ITGB3/2597] antibody to Integrin beta 3 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: ITGB3/2597
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human