Anti-Albumin Mouse Monoclonal Antibody [clone: ALB/2356]
ANTIA251684-100
New Product
- Antibody type:Primary
- Antigen name:alb
- Antigen symbol:Albumin
- Clonality:Monoclonal
- Clone:ALB/2356
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P02768
- Antigen synonyms:GIG20|DKFZp779N1935|ALBU_HUMAN|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Growth inhibiting protein 20|Albumin
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:66 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ALB protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [ALB/2356] antibody to Albumin for ELISA, Flow Cytometry, IF and WB with samples derived from Human.
- Validated applications: ELISA, Flow cytometry, IF, WB
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 2 to 4 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: Albumin
Clonality: Monoclonal
Clone: ALB/2356
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human