Order Entry
Export
ContactUsLinkComponent
Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: CDH16/2125]
Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: CDH16/2125]
Catalog # ANTIA251251-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cadherin 16 Mouse Monoclonal Antibody [clone: CDH16/2125]
Catalog # ANTIA251251-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CAD16_HUMAN
  • Antigen symbol:
    Cadherin 16
  • Clonality:
    Monoclonal
  • Clone:
    CDH16/2125
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O75309
  • Antigen synonyms:
    CDH16|Cadherin-16|CDH 16|Kidney specific cadherin|Cadherin 16 KSP cadherin|Cadherin 16|Kidney cadherin|Cadherin kidney|Kidney-specific cadherin
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    130 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 371 - 507, of human CDH16 protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CDH16/2125] antibody to Cadherin 16 for ELISA, WB and IHC-P with samples derived from Human.

  • Validated applications: ELISA, WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cadherin 16
Clonality: Monoclonal
Clone: CDH16/2125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human