Anti-Cytokeratin 5 + 6 + 18 Mouse Monoclonal Antibody [clone: LP34]
ANTIA250959-100
New Product
- Antibody type:Primary
- Antigen symbol:Cytokeratin 5 + 6 + 18
- Clonality:Monoclonal
- Clone:LP34
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine,Rat
- Form:Liquid
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:45 - 58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Detergent-insoluble fraction of psoriatic human epidermis
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [LP34] antibody to Cytokeratin 5 + 6 + 18 for IHC-P with samples derived from Human, Bovine and Rat.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 5 + 6 + 18
Clonality: Monoclonal
Clone: LP34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Bovine;Rat