Anti-Cytokeratin 7 + 17 Mouse Monoclonal Antibody [clone: C-46]
Catalog # ANTIA250880-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:Cytokeratin 7 + 17
- Clonality:Monoclonal
- Clone:C-46
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08729, Q04695
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:55 (CK7)/46 kDa (CK17)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeletal preparation from HeLa cells (human cervical carcinoma cells)
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-46] antibody to Cytokeratin 7 + 17 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human and Bovine.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 7 + 17
Clonality: Monoclonal
Clone: C-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Bovine