Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: 23C6]
Catalog # ANTIA250849-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:antigen identified by monoclonal
- Antigen symbol:Integrin alpha V
- Clonality:Monoclonal
- Clone:23C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Chicken,Human
- Form:Liquid
- Gene ID:UniprotID# P06756
- Antigen synonyms:Integrin alpha-5|CD 51|DKFZp686A08142|Integrin alpha-V light chain|CD51|integrin alpha V beta 3|Integrin alpha V|integrin alpha-V|Integrin alpha five
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:150 - 160 kDa under non - reducing conditions, Under reducing conditions, cleaved into 2 bands of about 125 - 130 kDa/20 - 25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Osteoclasts from osteoclastomas
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [23C6] antibody to Integrin alpha V for ELISA with samples derived from Human and Chicken.
- Validated applications: ELISA
Type: Primary
Antigen: Integrin alpha V
Clonality: Monoclonal
Clone: 23C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Chicken