Anti-CD59 Mouse Monoclonal Antibody [clone: MACIF/629]
Catalog # ANTIA250740-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:1F5 antigen
- Antigen symbol:CD59
- Clonality:Monoclonal
- Clone:MACIF/629
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P13987
- Antigen synonyms:CD59 antigen|CD59 antigen complement regulatory protein|1F5|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|16.3A5|20 kDa homologous restriction factor|CD59 antigen p18 20
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:20 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human K562 tumor cells
- Tested applications:Functional studies, IHC-P
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MACIF/629] antibody to CD59 for Functional Studies, Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated applications: Functional Studies, Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MACIF/629
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human