Order Entry
Export
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]
Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]
  ANTIA250736-100
New Product
undefined
Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]
  ANTIA250736-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Antigen CD48
  • Antigen symbol:
    CD48
  • Clonality:
    Monoclonal
  • Clone:
    5-4.8
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P09326
  • Antigen synonyms:
    BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human peripheral blood lymphocytes
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [5-4.8] antibody to CD48 for Flow Cytometry and IF with samples derived from Human.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 5-4.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human