Order Entry
Export
ContactUsLinkComponent
 
Anti-CD36 Mouse Monoclonal Antibody [clone: 185-1G2]
  ANTIA250686-100
New Product
undefined
Anti-CD36 Mouse Monoclonal Antibody [clone: 185-1G2]
  ANTIA250686-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Adipocyte membrane protein
  • Antigen symbol:
    CD36
  • Clonality:
    Monoclonal
  • Clone:
    185-1G2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16671
  • Antigen synonyms:
    CHDS7|CD36|BDPLT10|CD36 molecule (thrombospondin receptor)|CD36 molecule|Cluster determinant 36|CD36 antigen|CD36_HUMAN|CD36 antigen (collagen type I receptor, thrombospondin receptor)
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    80 - 90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Stimulated human leukocytes
  • Tested applications:
    Functional studies
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [185-1G2] antibody to CD36 for Functional Studies, Flow Cytometry and IF with samples derived from Human.

  • Validated applications: Functional Studies, Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 185-1G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human