Order Entry
Export
ContactUsLinkComponent
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEnd/10 + HPCA1/763]
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEnd/10 + HPCA1/763]
  ANTIA250680-100
New Product
undefined
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEnd/10 + HPCA1/763]
  ANTIA250680-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD34
  • Antigen symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    QBEnd/10 + HPCA1/763
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    Multiple
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28906
  • Antigen synonyms:
    CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    90 - 110 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Clone QBEnd/10: Detergent solubilised vesicular suspension prepared from human term placenta. Clone HPCA1/763: Recombinant full-length human HPCA1 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [QBEnd/10 + HPCA1/763] antibody to CD34 for Flow Cytometry, IF and IHC-P with samples derived from Human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd/10 + HPCA1/763
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human